ENA432537976 Enamine

4-(5-chloro-2-methoxyphenyl)-1H-1,2,3-triazole

  • MDL number MFCD28139478
  • Cas Number : 1803587-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193444
Availability details for
Quantity
  • Availability
  • Add to Cart