ENA432537985 Enamine

methyl 1-tert-butyl-2-(1-methyl-1H-pyrazol-4-yl)-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD28145242
  • Cas Number : 1803562-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193490
Availability details for
Quantity
  • Availability
  • Add to Cart