ENA432537987 Enamine

2-chloro-4-fluoropyridine-3-sulfonyl chloride

  • MDL number MFCD28139488
  • Cas Number : 1803567-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193503
Availability details for
Quantity
  • Availability
  • Add to Cart