ENA432537994 Enamine

2,3-dimethyl-1lambda4-thiomorpholin-1-one hydrochloride

  • MDL number MFCD28139495
  • Cas Number : 1803609-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-193594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193594
Availability details for
Quantity
  • Availability
  • Add to Cart