ENA432537998 Enamine

2-amino-2-[2-(trifluoromethoxy)phenyl]ethan-1-ol hydrochloride

  • MDL number MFCD28139500
  • Cas Number : 1803599-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193659
Availability details for
Quantity
  • Availability
  • Add to Cart