ENA432538011 Enamine

2-[4-(trimethylsilyl)phenyl]acetonitrile

  • MDL number MFCD28139521
  • Cas Number : 17983-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194186
Availability details for
Quantity
  • Availability
  • Add to Cart