ENA432538012 Enamine

3-(azetidin-3-yl)-1,2-oxazole hydrochloride

  • MDL number MFCD28139522
  • Cas Number : 1700260-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194191
Availability details for
Quantity
  • Availability
  • Add to Cart