ENA432538017 Enamine

3-amino-5-methyl-1,2-oxazole-4-sulfonyl chloride

  • MDL number MFCD28139530
  • Cas Number : 1701865-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194325
Availability details for
Quantity
  • Availability
  • Add to Cart