ENA432538029 Enamine

3-(3-aminophenyl)-5-ethylimidazolidine-2,4-dione hydrochloride

  • MDL number MFCD28139542
  • Cas Number : 1803562-16-4
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-aminophenyl)-5-ethylimidazolidine-2,4-dione hydrochloride
Inchi Code 1S/C11H13N3O2.ClH/c1-2-9-10(15)14(11(16)13-9)8-5-3-4-7(12)6-8;/h3-6,9H,2,12H2,1H3,(H,13,16);1H
Inchi Key RJPZRMZBFNOENV-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-194408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194408
Availability details for
Quantity
  • Availability
  • Add to Cart