ENA432538030 Enamine

tert-butyl N-{[(2,2-difluoroethyl)carbamoyl]methyl}carbamate

  • MDL number MFCD28139545
  • Cas Number : 1414642-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194430
Availability details for
Quantity
  • Availability
  • Add to Cart