ENA432538032 Enamine

3-(4-methoxypyridin-3-yl)-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD28139549
  • Cas Number : 1803610-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194597
Availability details for
Quantity
  • Availability
  • Add to Cart