ENA432538040 Enamine

4-(methoxymethyl)piperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD28139555
  • Cas Number : 1803581-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194756
Availability details for
Quantity
  • Availability
  • Add to Cart