ENA432538042 Enamine

1,3-diethyl 4-acetyl-2-(3-bromophenyl)-5-hydroxy-5-methylcyclohexane-1,3-dicarboxylate

  • MDL number MFCD28145253
  • Cas Number : 1803581-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194822
Availability details for
Quantity
  • Availability
  • Add to Cart