ENA432538057 Enamine

7-fluoro-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD15832811
  • Cas Number : 410086-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231-234
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112579
Availability details for
Quantity
  • Availability
  • Add to Cart