ENA432538062 Enamine

5-amino-3-(methylsulfanyl)-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD11208105
  • Cas Number : 1152621-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182351
Availability details for
Quantity
  • Availability
  • Add to Cart