ENA432538071 Enamine

3-methyl-1,2-thiazole-5-sulfonyl chloride

  • MDL number MFCD21362487
  • Cas Number : 1355334-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-183255
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart