ENA432538075 Enamine

ethyl 2-(3-methylpyridin-2-yl)-2-oxoacetate

  • MDL number MFCD11975565
  • Cas Number : 1260861-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184350
Availability details for
Quantity
  • Availability
  • Add to Cart