ENA432538077 Enamine

1-(2-hydroxyethyl)-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD28139371
  • Cas Number : 1795430-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184456
Availability details for
Quantity
  • Availability
  • Add to Cart