ENA432538080 Enamine

1-methyl-3-(propan-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD21666964
  • Cas Number : 1393101-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-185687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185687
Availability details for
Quantity
  • Availability
  • Add to Cart