ENA432538082 Enamine

5-chloro-3-methyl-1,2-oxazole

  • MDL number MFCD11216975
  • Cas Number : 1314938-49-2
  • Molecular Weight : 117.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-methylisoxazole
Inchi Code 1S/C4H4ClNO/c1-3-2-4(5)7-6-3/h2H,1H3
Inchi Key JKYWQVCSFFZMSB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114081

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-114081
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart