ENA432538092 Enamine

3-chloro-2-nitrobenzamide

  • MDL number MFCD11110986
  • Cas Number : 59772-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187833
Availability details for
Quantity
  • Availability
  • Add to Cart