ENA432538093 Enamine

3-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD13293244
  • Cas Number : 1292238-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187857
Availability details for
Quantity
  • Availability
  • Add to Cart