ENA432538097 Enamine

{5H,6H,8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-3-yl}methanol

  • MDL number MFCD28139374
  • Cas Number : 1798716-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188057
Availability details for
Quantity
  • Availability
  • Add to Cart