ENA432538100 Enamine

1-{5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-6-yl}ethan-1-one

  • MDL number MFCD08361931
  • Cas Number : 907971-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188218
Availability details for
Quantity
  • Availability
  • Add to Cart