ENA432538103 Enamine

1-tert-butyl-3-(propan-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11180124
  • Cas Number : 1152506-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188378
Availability details for
Quantity
  • Availability
  • Add to Cart