ENA432538110 Enamine

2-chloro-5-propanoylbenzene-1-sulfonyl chloride

  • MDL number MFCD10694279
  • Cas Number : 1152589-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188626
Availability details for
Quantity
  • Availability
  • Add to Cart