ENA432538112 Enamine

1-methyl-2-oxo-2,3-dihydro-1H-indole-6-carboxylic acid

  • MDL number MFCD17015858
  • Cas Number : 138328-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188808
Availability details for
Quantity
  • Availability
  • Add to Cart