ENA432538113 Enamine

2-(thiophen-2-yl)-2-[(trimethylsilyl)oxy]acetonitrile

  • MDL number MFCD16786603
  • Cas Number : 82069-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188828
Availability details for
Quantity
  • Availability
  • Add to Cart