ENA432538117 Enamine

4-(pyrrolidine-1-sulfonyl)pyridin-2-amine

  • MDL number MFCD16688501
  • Cas Number : 1247138-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189040
Availability details for
Quantity
  • Availability
  • Add to Cart