ENA432538121 Enamine

(1R,3S)-3-(propan-2-yl)-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD28390117
  • Cas Number : 1820580-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189195
Availability details for
Quantity
  • Availability
  • Add to Cart