ENA432538130 Enamine

2-(3,4-dimethoxy-2-methylphenyl)acetonitrile

  • MDL number MFCD27928343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189571
Availability details for
Quantity
  • Availability
  • Add to Cart