ENA432538131 Enamine

N-ethyl-N-methyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD12806209
  • Cas Number : 1178766-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189626
Availability details for
Quantity
  • Availability
  • Add to Cart