ENA432538139 Enamine

5-ethynylpyridine-3-carboxylic acid

  • MDL number MFCD18207405
  • Cas Number : 1211533-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-120575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120575
Availability details for
Quantity
  • Availability
  • Add to Cart