ENA432538145 Enamine

6-chloro-2-(4-fluorophenyl)-3-nitropyridine

  • MDL number MFCD22833113
  • Cas Number : 1521169-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189881
Availability details for
Quantity
  • Availability
  • Add to Cart