ENA432538147 Enamine

2-[4-(acetyloxy)phenyl]acetic acid

  • MDL number MFCD00033272
  • Cas Number : 38177-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189918
Availability details for
Quantity
  • Availability
  • Add to Cart