ENA432538148 Enamine

1-(3-bromophenyl)-3-chloropropan-2-one

  • MDL number MFCD11621966
  • Cas Number : 1095033-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189920
Availability details for
Quantity
  • Availability
  • Add to Cart