ENA432538150 Enamine

hexane-3-thiol

  • MDL number MFCD00053517
  • Cas Number : 1633-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189975
Availability details for
Quantity
  • Availability
  • Add to Cart