ENA432538157 Enamine

3-bromo-3-methylbutan-2-one

  • MDL number MFCD06661470
  • Cas Number : 2648-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121902

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P264,P280,P302+P352,P303+P361+P353,P305+P351+P338,P332+P313,P337+P313,P362,P370+P378,P403+P235,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-121902
pictograms GHS02,GHS07
hstatements H226,H315,H319
Availability details for
Quantity
  • Availability
  • Add to Cart