ENA432538158 Enamine

[4-chloro-2-(1H-1,2,4-triazol-1-yl)phenyl]methanol

  • MDL number MFCD28139383
  • Cas Number : 1597924-54-3
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chloro-2-(1H-1,2,4-triazol-1-yl)phenyl)methanol
Inchi Code 1S/C9H8ClN3O/c10-8-2-1-7(4-14)9(3-8)13-6-11-5-12-13/h1-3,5-6,14H,4H2
Inchi Key UEHMNGRIRJPSQS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-190326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190326
Availability details for
Quantity
  • Availability
  • Add to Cart