ENA432538164 Enamine

ethyl 1-(3-cyanophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD28139385
  • Cas Number : 1027551-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190411
Availability details for
Quantity
  • Availability
  • Add to Cart