ENA432538166 Enamine

(2S)-2-(naphthalen-2-yloxy)propanoic acid

  • MDL number MFCD24479576
  • Cas Number : 25258-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190513
Availability details for
Quantity
  • Availability
  • Add to Cart