ENA432538181 Enamine

4-(4-iodophenyl)butan-2-one

  • MDL number MFCD21243165
  • Cas Number : 918540-55-3
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 74-78
Iupac Name 4-(4-iodophenyl)-2-butanone
Inchi Code 1S/C10H11IO/c1-8(12)2-3-9-4-6-10(11)7-5-9/h4-7H,2-3H2,1H3
Inchi Key KRODAABNJFTHLD-UHFFFAOYSA-N

Description

Melting Point 74-78
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-190846

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-190846
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart