ENA432538182 Enamine

tert-butyl N-{3-hydroxy-2-[(1-methyl-1H-pyrazol-5-yl)methyl]propyl}carbamate

  • MDL number MFCD28145229
  • Cas Number : 1803607-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190866
Availability details for
Quantity
  • Availability
  • Add to Cart