ENA432538187 Enamine

2-(1,2,3,4-tetrahydronaphthalen-1-yloxy)acetic acid

  • MDL number MFCD09929360
  • Cas Number : 120681-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191027
Availability details for
Quantity
  • Availability
  • Add to Cart