ENA432538205 Enamine

2-(6-ethoxypyridin-2-yl)acetic acid

  • MDL number MFCD26912407
  • Cas Number : 1550789-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191345
Availability details for
Quantity
  • Availability
  • Add to Cart