ENA432538214 Enamine

4-hydrazinyl-1-methyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD26046797
  • Cas Number : 1565576-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-128328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128328
Availability details for
Quantity
  • Availability
  • Add to Cart