ENA432538217 Enamine

tert-butyl N-[1-(hydroxymethyl)cyclobutyl]carbamate

  • MDL number MFCD12028429
  • Cas Number : 1142211-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131146
Availability details for
Quantity
  • Availability
  • Add to Cart