ENA432538219 Enamine

methyl 3-hydroxy-5-nitrobenzoate

  • MDL number MFCD00463877
  • Cas Number : 55076-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191536
Availability details for
Quantity
  • Availability
  • Add to Cart