ENA432538244 Enamine

1-(2,4-difluorophenyl)-2-methylpropan-1-one

  • MDL number MFCD11104172
  • Cas Number : 151856-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134154
Availability details for
Quantity
  • Availability
  • Add to Cart