ENA432538247 Enamine

1-chloro-5-methylhexan-2-ol

  • MDL number MFCD19232587
  • Cas Number : 107323-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191975
Availability details for
Quantity
  • Availability
  • Add to Cart